missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) oxide, 99%, Thermo Scientific Chemicals

36.08 € - 479.39 €

Chemical Identifiers

CAS 21651-19-4
Molecular Formula OSn
Molecular Weight (g/mol) 134.71
MDL Number MFCD00011243
InChI Key QHGNHLZPVBIIPX-UHFFFAOYSA-N
Synonym stannous oxide, tin ii oxide, tin oxide sno, tin monoxide, tin oxide sn2o2, stannane, oxo, stannanone, stannicoxide, oxostannanylidene, oxo stannane
PubChem CID 88989
IUPAC Name oxotin
SMILES O=[Sn]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11463498
SDS View Documents
Thermo Scientific Alfa Aesar
A17889.22
100 g
36.08 €
100g
Please to purchase this item. Need a web account? Register with us today!
11473498
SDS View Documents
Thermo Scientific Alfa Aesar
A17889.36
500 g
124.62 €
500g
Please to purchase this item. Need a web account? Register with us today!
11483498
SDS View Documents
Thermo Scientific Alfa Aesar
A17889.0E
2500 g
479.39 €
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tin(II) oxide is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tin(II) oxide is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.

Solubility
Insoluble in water and alcohol.

Notes
Incompatible with materials like strong oxidizing agents, strong acids, strong reducing agents and alkali metals.
Specifications

Chemical Identifiers

21651-19-4
134.71
QHGNHLZPVBIIPX-UHFFFAOYSA-N
88989
O=[Sn]
OSn
MFCD00011243
stannous oxide, tin ii oxide, tin oxide sno, tin monoxide, tin oxide sn2o2, stannane, oxo, stannanone, stannicoxide, oxostannanylidene, oxo stannane
oxotin

Specifications

1080°C (decomposition)
OSn
MFCD00011243
stannous oxide, tin ii oxide, tin oxide sno, tin monoxide, tin oxide sn2o2, stannane, oxo, stannanone, stannicoxide, oxostannanylidene, oxo stannane
QHGNHLZPVBIIPX-UHFFFAOYSA-N
oxotin
88989
99%
6.45
21651-19-4
SnO
14,8787
Insoluble in water and alcohol.
O=[Sn]
134.71
134.69
Odorless
Tin(II) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P260-P271-P304+P340-P312-P314-P501c

H335-H373

missing translation for 'einecsNumber' : 244-499-5

missing translation for 'rtecsNumber' : XQ3700000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(II) oxide, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.