missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) iodide, 99+%, Thermo Scientific Chemicals

138.69 € - 516.57 €

Chemical Identifiers

CAS 10294-70-9
Molecular Formula I2Sn
Molecular Weight (g/mol) 372.519
MDL Number MFCD00049545
InChI Key JTDNNCYXCFHBGG-UHFFFAOYSA-L
Synonym tin ii iodide, stannous iodide, tin diiodide, tin iodide sni2, stannous diiodide, sni2, acmc-1bqot, tin ii iodide, ultra dry, tin ii iodide,-10 mesh trace metals basis, tin ii iodide, anhydrous, beads,-10 mesh trace metals basis
PubChem CID 25138
IUPAC Name diiodotin
SMILES [Sn](I)I
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11314809
View Documents
Thermo Scientific Alfa Aesar
071112.09
10 g
138.69 €
10g
Please to purchase this item. Need a web account? Register with us today!
11304809
View Documents
Thermo Scientific Alfa Aesar
071112.18
50 g
516.57 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tin(II) iodide is used to prepare organic-inorganic perovskite thin films which exhibit uniform surfaces and strong photoluminescence. It finds application as an analytical reagent in the determination of rhodium and iridium by spectrophotometry. It is also involved in the synthesis of CH3(CH2)11NH3]SnI3, which is a hybrid semiconductor with MoO3 type tin(II) iodide layers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10294-70-9
372.519
JTDNNCYXCFHBGG-UHFFFAOYSA-L
25138
[Sn](I)I
I2Sn
MFCD00049545
tin ii iodide, stannous iodide, tin diiodide, tin iodide sni2, stannous diiodide, sni2, acmc-1bqot, tin ii iodide, ultra dry, tin ii iodide,-10 mesh trace metals basis, tin ii iodide, anhydrous, beads,-10 mesh trace metals basis
diiodotin

Specifications

720°C (decomposition)
10294-70-9
>99%
MFCD00049545
Soluble in benzene,carbon disulfide,chloroform,and solutions of alkali chlorides and iodides. Decomposes in water
[Sn](I)I
372.519
372.5
5.28
320°C
Powder
I2Sn
tin ii iodide, stannous iodide, tin diiodide, tin iodide sni2, stannous diiodide, sni2, acmc-1bqot, tin ii iodide, ultra dry, tin ii iodide,-10 mesh trace metals basis, tin ii iodide, anhydrous, beads,-10 mesh trace metals basis
JTDNNCYXCFHBGG-UHFFFAOYSA-L
diiodotin
25138
Air, light, and moisture sensitive
Tin(II) iodide
Videos
Safety and Handling

Safety and Handling

When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : DOT Class: 8; Packaging Group: III

missing translation for 'einecsNumber' : 233-667-3

missing translation for 'rtecsNumber' : XQ3650000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only