Learn More
Tin(II) chloride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
69.45 € - 815.06 €
Chemical Identifiers
CAS | 1370709-86-6 |
---|---|
Molecular Formula | Cl2Sn |
Molecular Weight (g/mol) | 189.61 |
MDL Number | MFCD00011241 |
InChI Key | AXZWODMDQAVCJE-UHFFFAOYSA-L |
Synonym | Stannous chloride hydrate |
PubChem CID | 24479 |
ChEBI | CHEBI:78067 |
SMILES | [Cl-].[Cl-].[Sn++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337067
|
Thermo Scientific Alfa Aesar
010894.06 |
5 g |
69.45 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327067
|
Thermo Scientific Alfa Aesar
010894.14 |
25 g |
266.33 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347067
|
Thermo Scientific Alfa Aesar
010894.22 |
100 g |
815.06 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It can be used to reduce certain metal salts into metals, tin plating, analytical testing of mercury ions, formation of silver mirrors and metal-metal bonds. It is also used for tin-plating, as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. Further it is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1370709-86-6 | |
189.61 | |
AXZWODMDQAVCJE-UHFFFAOYSA-L | |
24479 | |
[Cl-].[Cl-].[Sn++] |
Cl2Sn | |
MFCD00011241 | |
Stannous chloride hydrate | |
CHEBI:78067 |
Specifications
43°C to 46°C | |
White | |
Cl2Sn | |
MFCD00011241 | |
14,8783 | |
Soluble in ethanol,acetone,ether and tetrahydrofuran. Insoluble in xylene. | |
[Cl-].[Cl-].[Sn++] | |
189.61 | |
CHEBI:78067 | |
99.995% | |
Odorless | |
2.71 g/mL | |
Tin(II) chloride hydrate |
1370709-86-6 | |
Crystalline Powder | |
SnCl2·xH2O | |
UN3260 | |
Stannous chloride hydrate | |
AXZWODMDQAVCJE-UHFFFAOYSA-L | |
dichlorotin | |
24479 | |
189.62 (Anhydrous) | |
Puratronic™ | |
Air Sensitive | |
(metals basis) |
Safety and Handling
GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P390-P501c
H290-H302+H332-H314-H317-H335-H373
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 231-868-0
missing translation for 'rtecsNumber' : XP8850000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only