missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride dihydrate, Reagent Grade, Thermo Scientific Chemicals

28.30 € - 357.78 €

Chemical Identifiers

CAS 10025-69-1
Molecular Formula SnCl2·2H2O
MDL Number MFCD00149863
Synonym Stannous chloride dihydrate
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11357227
SDS View Documents
Thermo Scientific Alfa Aesar
011536.09
10 g N/A N/A N/A
11337227
SDS View Documents
Thermo Scientific Alfa Aesar
011536.22
100 g
40.90 €
100g
Please to purchase this item. Need a web account? Register with us today!
11367227
SDS View Documents
Thermo Scientific Alfa Aesar
011536.36
500 g
120.60 €
500g
Please to purchase this item. Need a web account? Register with us today!
11347227
SDS View Documents
Thermo Scientific Alfa Aesar
011536.A3
2 kg
357.78 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Stannous chloride is used as a sensitizing agent in preparing glass and plastic for metalizing. It serves as a potent reducing agent. Other uses of stannous chloride are tin electroplating baths, corrosion inhibitors, polymers, thermoplastic elastomers, soldering flux, antioxidant, tanning agent and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10025-69-1
SnCl2·2H2O
MFCD00149863
Stannous chloride dihydrate

Specifications

10025-69-1
SnCl2·2H2O
Stannous chloride dihydrate
Tin(II) chloride dihydrate, Reagent Grade
Crystalline
MFCD00149863
225.65 (189.62 Anhydrous)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-868-0

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(II) chloride dihydrate, Reagent Grade, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.