missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) Chloride Dihydrate, 98%, ACS Reagent, Honeywell Fluka™

Product Code. 15642620
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
12 kg
Packaging:
Drum with inliner
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent, 98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-69-1
MDL Number MFCD00149863

Specifications

Boiling Point 652°C
Melting Point 37°C to 38°C
Color Colorless
Quantity 100 g
Linear Formula SnCl2 · 2H2O
UN Number UN3260
Solubility Information Soluble in water, Hydrochloric Acid
Formula Weight 225.65g/mol
Percent Purity 98%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Crystalline
Density 2.71g/cm3
Chemical Name or Material Tin(II) Chloride Dihydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tin(II) chloride dihydrate
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'