missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymol Blue

35.28 € - 317.58 €

Chemical Identifiers

CAS 76-61-9
Molecular Formula C27H30O5S
Molecular Weight (g/mol) 466.592
MDL Number MFCD00005869
InChI Key PRZSXZWFJHEZBJ-UHFFFAOYSA-N
Synonym TB, Thymolsulfonephthalein
PubChem CID 65565
IUPAC Name 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10331311
SDS View Documents
Thermo Scientific Chemicals
420930050
5 g Glass bottle
35.28 €
5g
Please to purchase this item. Need a web account? Register with us today!
10697401
SDS View Documents
Thermo Scientific Chemicals
420930100
10 g Glass bottle
65.93 €
10g
Please to purchase this item. Need a web account? Register with us today!
10288903
SDS View Documents
Thermo Scientific Chemicals
420931000
100 g Glass bottle
317.58 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

76-61-9
466.592
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
65565
CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
C27H30O5S
MFCD00005869
TB, Thymolsulfonephthalein
4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol

Specifications

76-61-9
C27H30O5S
15, 9555
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O
466.592
466.58
Acid range from pH 1.2 (pink) to pH 2.8 (yellow), Alkaline range from pH 8.0 (yellow) to pH 9.2 (blue)
Fine Crystalline Powder
Passes Test
MFCD00005869
TB, Thymolsulfonephthalein
Authentic
4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
65565
ACS Reagent
Brown to Green
Thymol Blue, For analysis ACS
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Thymol Blue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.