Learn More
Thulium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
109.55 € - 409.04 €
Chemical Identifiers
CAS | 141478-68-4 |
---|---|
Molecular Formula | C3F9O9S3Tm |
Molecular Weight (g/mol) | 616.124 |
MDL Number | MFCD00209617 |
InChI Key | PBASUZORNBYVFM-UHFFFAOYSA-K |
Synonym | thulium iii trifluoromethanesulfonate, thulium iii triflate, trifluoromethanesulfonic acid thulium iii salt, thulium triflate, tm otf 3, acmc-1c8m6, thulium 3+ tritriflate, thulium 3+ ; trifluoromethanesulfonate, thulium trifluoromethanesulfonate, thulium iii tris trifluoromethanesulfonate |
PubChem CID | 2733936 |
IUPAC Name | thulium(3+);trifluoromethanesulfonate |
SMILES | C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Tm+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11363868
|
Thermo Scientific Alfa Aesar
040313.04 |
2 g |
109.55 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11373868
|
Thermo Scientific Alfa Aesar
040313.09 |
10 g |
409.04 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A raw material used in synthesis of chemicals .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141478-68-4 | |
616.124 | |
PBASUZORNBYVFM-UHFFFAOYSA-K | |
2733936 | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Tm+3] |
C3F9O9S3Tm | |
MFCD00209617 | |
thulium iii trifluoromethanesulfonate, thulium iii triflate, trifluoromethanesulfonic acid thulium iii salt, thulium triflate, tm otf 3, acmc-1c8m6, thulium 3+ tritriflate, thulium 3+ ; trifluoromethanesulfonate, thulium trifluoromethanesulfonate, thulium iii tris trifluoromethanesulfonate | |
thulium(3+);trifluoromethanesulfonate |
Specifications
141478-68-4 | |
C3F9O9S3Tm | |
MFCD00209617 | |
Soluble in water. | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Tm+3] | |
616.124 | |
616.13 | |
Thulium(III) trifluoromethanesulfonate |
Powder | |
Tm(OSO2CF3)3 | |
thulium iii trifluoromethanesulfonate, thulium iii triflate, trifluoromethanesulfonic acid thulium iii salt, thulium triflate, tm otf 3, acmc-1c8m6, thulium 3+ tritriflate, thulium 3+ ; trifluoromethanesulfonate, thulium trifluoromethanesulfonate, thulium iii tris trifluoromethanesulfonate | |
PBASUZORNBYVFM-UHFFFAOYSA-K | |
thulium(3+);trifluoromethanesulfonate | |
2733936 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only