Learn More
Thulium(III) sulfate octahydrate, 99.99%, Thermo Scientific™
42.91 € - 142.00 €
Chemical Identifiers
CAS | 13778-40-0 |
---|---|
Molecular Formula | H16O20S3Tm2 |
Molecular Weight (g/mol) | 770.156 |
MDL Number | MFCD00149881 |
InChI Key | GPBYLGMJMIXKMU-UHFFFAOYSA-H |
Synonym | thulium sulfate octahydrate, unii-8zgz55l20j, thulium sulfate octahydrate mi, acmc-20aji8, thulium sulfate tm2 so4 3 octahydrate, thulium sulfate hydrate tm2 so4 3.8h2o, sulfuric acid, thulium 3+ salt 3:2 , octahydrate, thulium sulfate-water 2/3/8, thulium sulfate,, sulfuric acid thulium iii salt octahydrate |
PubChem CID | 71310074 |
IUPAC Name | thulium(3+);trisulfate;octahydrate |
SMILES | O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Tm+3].[Tm+3] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used as a reactant in dehydration reactions. It is the important dopant for fiber amplifiers.
Solubility
Soluble in water, moderately soluble in strong mineral acids.
Notes
Store at room temperature. Keep away from strong oxidizing agents and moisture.
Chemical Identifiers
13778-40-0 | |
770.156 | |
GPBYLGMJMIXKMU-UHFFFAOYSA-H | |
71310074 | |
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Tm+3].[Tm+3] |
H16O20S3Tm2 | |
MFCD00149881 | |
thulium sulfate octahydrate, unii-8zgz55l20j, thulium sulfate octahydrate mi, acmc-20aji8, thulium sulfate tm2 so4 3 octahydrate, thulium sulfate hydrate tm2 so4 3.8h2o, sulfuric acid, thulium 3+ salt 3:2 , octahydrate, thulium sulfate-water 2/3/8, thulium sulfate,, sulfuric acid thulium iii salt octahydrate | |
thulium(3+);trisulfate;octahydrate |
Specifications
13778-40-0 | |
MFCD00149881 | |
Soluble in water,moderately soluble in strong mineral acids. | |
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Tm+3].[Tm+3] | |
770.156 | |
770.17 (626.06 Anhydrous) | |
Hygroscopic |
H16O20S3Tm2 | |
thulium sulfate octahydrate, unii-8zgz55l20j, thulium sulfate octahydrate mi, acmc-20aji8, thulium sulfate tm2 so4 3 octahydrate, thulium sulfate hydrate tm2 so4 3.8h2o, sulfuric acid, thulium 3+ salt 3:2 , octahydrate, thulium sulfate-water 2/3/8, thulium sulfate,, sulfuric acid thulium iii salt octahydrate | |
GPBYLGMJMIXKMU-UHFFFAOYSA-H | |
thulium(3+);trisulfate;octahydrate | |
71310074 | |
99.99% | |
Thulium(III) sulfate octahydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 243-996-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.