missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) fluoride, ultra dry, 99.99% (REO), Thermo Scientific™

107.00 € - 497.00 €

Chemical Identifiers

CAS 13760-79-7
Molecular Formula F3Tm
Molecular Weight (g/mol) 225.93
MDL Number MFCD00049611
InChI Key AATUHDXSJTXIHB-UHFFFAOYSA-K
IUPAC Name thulium(3+) trifluoride
SMILES [F-].[F-].[F-].[Tm+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11358937
SDS View Documents
Thermo Scientific Alfa Aesar
038615.03
1 g N/A N/A N/A
11348937
SDS View Documents
Thermo Scientific Alfa Aesar
038615.06
5 g N/A N/A N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
Specifications

Chemical Identifiers

13760-79-7
225.93
AATUHDXSJTXIHB-UHFFFAOYSA-K
[F-].[F-].[F-].[Tm+3]
F3Tm
MFCD00049611
thulium(3+) trifluoride

Specifications

2200°C
13760-79-7
F3Tm
UN3288
AATUHDXSJTXIHB-UHFFFAOYSA-K
thulium(3+) trifluoride
225.93
Odorless
Hygroscopic
(REO)
1158°C
Powder
MFCD00049611
It is insoluble in water.
[F-].[F-].[F-].[Tm+3]
225.93
99.99%
Ampule
7.9 g/mL
Thulium(III) fluoride, Ultra dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 237-353-7

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thulium(III) fluoride, ultra dry, 99.99% (REO), Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.