Learn More
Thulium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
134.67 € - 1690.00 €
Chemical Identifiers
CAS | 314041-04-8 |
---|---|
Molecular Formula | C6H9O6Tm |
Molecular Weight (g/mol) | 346.07 |
MDL Number | MFCD00013046 |
InChI Key | SNMVVAHJCCXTQR-UHFFFAOYSA-K |
Synonym | acmc-1afih, thulium acetate-water 1/3/1, thulium 3+ ion triacetate hydrate |
IUPAC Name | thulium(3+) triacetate |
SMILES | [Tm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11312538
|
Thermo Scientific Alfa Aesar
014582.04 |
2 g |
134.67 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11302538
|
Thermo Scientific Alfa Aesar
014582.09 |
10 g |
478.38 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11440470
|
Thermo Scientific Alfa Aesar
014582.18 |
50 g | N/A | N/A | N/A | |||||
Description
Thulium Acetate is the important dopant for silica-based fiber amplifiers. It can also be used in portable X-ray devices that has been bombarded in a nuclear reactor as a radiation source. Used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. The other main application is used for optical fiber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
314041-04-8 | |
346.07 | |
SNMVVAHJCCXTQR-UHFFFAOYSA-K | |
thulium(3+) triacetate |
C6H9O6Tm | |
MFCD00013046 | |
acmc-1afih, thulium acetate-water 1/3/1, thulium 3+ ion triacetate hydrate | |
[Tm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
Specifications
314041-04-8 | |
C6H9O6Tm | |
acmc-1afih, thulium acetate-water 1/3/1, thulium 3+ ion triacetate hydrate | |
SNMVVAHJCCXTQR-UHFFFAOYSA-K | |
thulium(3+) triacetate | |
346.07 (Anhydrous) | |
Reacton™ | |
(REO) |
Crystalline Aggregates | |
MFCD00013046 | |
Moderately soluble in water,strong mineral acids. | |
[Tm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O | |
346.07 | |
99.9% | |
Hygroscopic | |
Thulium(III) acetate hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 254-326-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.