Learn More
THRAP3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19744-1-AP-20UL
 View more versions of this product
Product Code. 16879224
  • 145.00 € / 20µL
Explore more special offers
Description
Description
The antibody recognizes the C-term of THRAP3.
THRAP3 is a 955 amino acid novel nuclear receptor and a member of the Thyroid hormone receptor associated proteins with a LISH domain and no obvious known sequence motifs. THRAP3 is a 150 kDa subunit of the large multiprotein complex TRAP and is important in the maintece of the fundamental structure of the fibrillar center and dense fibrillar component in the nucleolus. It is also known to play an important role in transcriptional coactivation. It is known to interact with DKC1, nop5/nap65 and fibrillarin. From the Northern blot analysis of several human tissues it was found that TRAP150 is ubiquitously expressed.
Specifications
Specifications
THRAP3
Polyclonal
Unconjugated
THRAP3
THRAP3, TRAP150
Rabbit
Antigen Affinity Chromatography
RUO
9967
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9Y2W1
THRAP3
Peptide
20 μL