Learn More
THOC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10920-1-AP-150UL
 View more versions of this product
Product Code. 16897623
  • 410.00 € / 150µL
Explore more special offers
Description
Description
This antibody recognizes the 84 kDa THOC1 protein.
TCEB1 is known as protein elongin C, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation. Western blots using two different antibodies (P100962-T200 and P100963_P050 / P100963_P100) against two unique regions of this protein target confirm the same apparent molecular weight in our tests.
Specifications
Specifications
THOC1
Polyclonal
Unconjugated
THOC1
HPR1, hTREX84, Nuclear matrix protein p84, P84, P84N5, THO complex 1, THO complex subunit 1, Tho1, THOC1
Rabbit
Antigen Affinity Chromatography
RUO
225160, 291797, 9984
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P59924, Q8R3N6, Q96FV9
Thoc1
THOC1 Fusion Protein Ag1352
150 μL
Primary
Human, Mouse, Rat