missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-carbonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11347906
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1641-09-4
Molecular Formula C5H3NS
Molecular Weight (g/mol) 109.146
MDL Number MFCD00151852
InChI Key GSXCEVHRIVLFJV-UHFFFAOYSA-N
Synonym 3-cyanothiophene, 3-thiophenecarbonitrile, thiophene-3-nitrile, pubchem5508, pubchem12351, acmc-209dqk, ksc176g4h
PubChem CID 74231
IUPAC Name thiophene-3-carbonitrile
SMILES C1=CSC=C1C#N

Specifications

Density 1.2
Boiling Point 205°C
Flash Point 86°C (186°F)
Odor Recognizable
Refractive Index 1.5615
Quantity 25 g
UN Number UN3276
Beilstein 107104
Formula Weight 109.15
Percent Purity 98%
Chemical Name or Material Thiophene-3-carbonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'