Learn More
Thiomorpholine, 98%, Thermo Scientific Chemicals
54.90 € - 695.00 €
Chemical Identifiers
CAS | 123-90-0 |
---|---|
Molecular Formula | C4H9NS |
Molecular Weight (g/mol) | 103.183 |
MDL Number | MFCD00005974 |
InChI Key | BRNULMACUQOKMR-UHFFFAOYSA-N |
Synonym | thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine |
PubChem CID | 67164 |
ChEBI | CHEBI:36392 |
IUPAC Name | thiomorpholine |
SMILES | C1CSCCN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11458516
|
Thermo Scientific Alfa Aesar
A15958.06 |
5 g |
54.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468516
|
Thermo Scientific Alfa Aesar
A15958.14 |
25 g |
190.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478516
|
Thermo Scientific Alfa Aesar
A15958.22 |
100 g |
695.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thiomorpholine is used as an intermediate for blonanserin. It is used in the preparation of pyrrole derivatives, which acts as a antimycobacterial agent. It is further involved in the hypocholesterolemic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThiomorpholine is used as an intermediate for blonanserin. It is used in the preparation of pyrrole derivatives, which acts as a antimycobacterial agent. It is further involved in the hypocholesterolemic activity.
Solubility
Miscible with waterMiscible with water and organic solvents.
Notes
Air sensitive. Incompatible with strong oxidizing agents, strong bases and air.
Chemical Identifiers
123-90-0 | |
103.183 | |
BRNULMACUQOKMR-UHFFFAOYSA-N | |
67164 | |
thiomorpholine |
C4H9NS | |
MFCD00005974 | |
thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine | |
CHEBI:36392 | |
C1CSCCN1 |
Specifications
123-90-0 | |
175°C to 177°C | |
C4H9NS | |
MFCD00005974 | |
102550 | |
14,9300 | |
Miscible with waterMiscible with water and organic solvents. | |
C1CSCCN1 | |
103.183 | |
CHEBI:36392 | |
98% |
1.086 | |
67°C (153°F) | |
1.5384 | |
UN3267 | |
Air Sensitive | |
thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine | |
BRNULMACUQOKMR-UHFFFAOYSA-N | |
thiomorpholine | |
67164 | |
103.19 | |
Thiomorpholine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 204-660-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only