Learn More
Thiobenzamide, 95%, Thermo Scientific Chemicals
26.13 € - 64.72 €
Chemical Identifiers
CAS | 2227-79-4 |
---|---|
Molecular Formula | C7H7NS |
Molecular Weight (g/mol) | 137.20 |
MDL Number | MFCD00008060 |
InChI Key | QIOZLISABUUKJY-UHFFFAOYSA-N |
Synonym | thiobenzamide, benzothioamide, benzamide, thio, benzothiamide, tiobenzamide, phenylthioamide, tiobenzamide italian, unii-8799vm9sxc, benzene-1-carbothioamide, aminophenylmethane-1-thione |
PubChem CID | 683563 |
ChEBI | CHEBI:80418 |
IUPAC Name | benzenecarbothioamide |
SMILES | NC(=S)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10113990
|
Thermo Scientific
164730100 |
10 g |
26.13 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10781241
|
Thermo Scientific Acros
164730500 |
50 g |
64.72 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2227-79-4 | |
137.20 | |
QIOZLISABUUKJY-UHFFFAOYSA-N | |
683563 | |
benzenecarbothioamide |
C7H7NS | |
MFCD00008060 | |
thiobenzamide, benzothioamide, benzamide, thio, benzothiamide, tiobenzamide, phenylthioamide, tiobenzamide italian, unii-8799vm9sxc, benzene-1-carbothioamide, aminophenylmethane-1-thione | |
CHEBI:80418 | |
NC(=S)C1=CC=CC=C1 |
Specifications
2227-79-4 | |
Yellow to Green | |
94% min. (HPLC) | |
C7H7NS | |
MFCD00008060 | |
thiobenzamide, benzothioamide, benzamide, thio, benzothiamide, tiobenzamide, phenylthioamide, tiobenzamide italian, unii-8799vm9sxc, benzene-1-carbothioamide, aminophenylmethane-1-thione | |
QIOZLISABUUKJY-UHFFFAOYSA-N | |
benzenecarbothioamide | |
683563 | |
137.21 | |
Crystalline Powder |
114°C to 118°C | |
Authentic | |
Glass bottle | |
C6H5CSNH2 | |
09, 424 | |
Solubility in water: slightly soluble | |
NC(=S)C1=CC=CC=C1 | |
137.20 | |
CHEBI:80418 | |
95% | |
Thiobenzamide, 95% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
Harmful if inhaled.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 218-765-6
missing translation for 'rtecsNumber' : CV5860000
missing translation for 'tsca' : TSCA
- Thiobenzamide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only