Learn More
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
45.10 € - 473.00 €
Chemical Identifiers
CAS | 62-55-5 |
---|---|
Molecular Formula | C2H5NS |
Molecular Weight (g/mol) | 75.13 |
MDL Number | MFCD00008070 |
InChI Key | YUKQRDCYNOVPGJ-UHFFFAOYSA-N |
Synonym | thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide |
PubChem CID | 2723949 |
ChEBI | CHEBI:32497 |
IUPAC Name | ethanethioamide |
SMILES | CC(N)=S |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10269610
|
Thermo Scientific Acros
424530250 |
25 g | Glass bottle |
45.10 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10617771
|
Thermo Scientific Acros
424531000 |
100 g | Glass bottle |
124.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10044013
|
Thermo Scientific Acros
424535000 |
500 g | Glass bottle |
473.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62-55-5 | |
75.13 | |
YUKQRDCYNOVPGJ-UHFFFAOYSA-N | |
2723949 | |
ethanethioamide |
C2H5NS | |
MFCD00008070 | |
thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide | |
CHEBI:32497 | |
CC(N)=S |
Specifications
62-55-5 | |
White to Beige | |
0.05% max. | |
99% min. (Argentometry) | |
CH3CSNH2 | |
02, 232 | |
15, 9472 | |
thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide | |
YUKQRDCYNOVPGJ-UHFFFAOYSA-N | |
ethanethioamide | |
2723949 | |
75.13 | |
99+% | |
Crystalline Powder |
112.0°C to 114.0°C | |
1.3700g/mL | |
Authentic | |
C2H5NS | |
MFCD00008070 | |
04,502 | |
1.37 | |
Solubility in water: 163g/L (25°C). Other solubilities: 26.4g/100g ethanol, sparingly soluble in ether, soluble in benzene | |
CC(N)=S | |
75.13 | |
CHEBI:32497 | |
(2% soln.) Passes test | |
ACS Reagent | |
Thioacetamide |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause cancer.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-541-4
- Thioacetamide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only