Learn More
Epredia™ Shandon™ Tissue-Loc™ Tissue Cassettes IV
Compatible with Epredia™ Shandon™ MicroWriter™ and PrintMate™ cassette labelers
Brand: Epredia™ 41728
This item is not returnable.
View return policy
Description
- Disposable
- Holds tissue specimens during embedding, processing
- Pore style: Small slots
- Supplied in box of 250 cassettes with attached lids
Specifications
Small Slots | |
Hinged | |
Lilac |