missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Mucolexx™
81.00 € - 800.00 €
Specifications
Color | Light Red |
---|---|
Product Type | Tissue Preservative |
For Use With (Application) | Provides short term preservation of specimens |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12648016
|
Epredia™
9990370 |
4 x 500mL (0.13 gal.) |
81.00 €
Pack of 4 |
In Stock
Log in to see stock. |
|||||
10473770
|
Epredia™
9990371 |
4 x 3.8L (1 gal.) |
89.20 €
Each |
In Stock
Log in to see stock. |
|||||
12560047
|
Epredia™
9990375 |
50 x 120mL, filled to 60mL |
800.00 €
Pack of 50 |
N/A | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
- Ready-to-use mucoliquefying preservative
- Liquefies (with agitation) mucoid cytology specimens
- Maintains cellular detail
- Short-term preservation of specimens
- Histology
- Cytology
Specifications
Light Red | |
Tissue Preservative | |
Provides short term preservation of specimens | |
Liquid |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H371-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P235-Keep cool.
- P314-Get medical advice/attention if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethyl alcohol, methyl alcohol, formaldehyde, sodium phosphate dibasic, sodium phosphate monobasic, l
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title