Learn More
Epredia™ Cassette 1 Biopsy Cassettes
Specially designed to hold biopsy specimens during processing and embedding
Brand:  Epredia™ 1000968
 View more versions of this product
Product Code. 15584815
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Made of acetal polymer
  • Cassette writing face is at a 30° angle
  • Pore style is small square
  • Locking lid attached by two hinges
  • Supplied in a box of 500 cassettes with attached breakaway lids
Order Info
Not available in Tube Packs
Specifications
Specifications
Square
Hinged
Acetal Polymer
30°
White
Biopsy Cassette