missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Trak 300™ Oil for the HEMATRAK™ system
Intended for use with the HEMATRAK™ automated differential system
Brand: Epredia™ M8000
59.25 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Oil does not contain PCBs and is safe to use with valuable optics
- Index of refraction is 1.5150 and viscosity is 300 cs
Specifications
Trak 300 Oil | |
For use with the Hematrak automated differential system | |
16 oz. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 473ml Trak 300 HEMATRAK Immersion Oil
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Terphenyl, hydrogenated, Isobutylene/butene copolymer
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction