Learn More
Thermo Scientific™ Fluophase™ RP Javelin™ Guard Columns
Offer convenient, easy to use protection for analytical HPLC columns
Brand: Thermo Scientific™ 82505-012106
This item is not returnable.
View return policy
Description
- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
- Perfluorinated phases provide unique selectivity
- Extra retention and selectivity for halogenenated compounds
- Excellent for taxane analysis
Specifications
5 μm | |
10 mm | |
2.1 mm | |
RP |
Product Content Correction
Product Title
Thermo Scientific™ Fluophase™ RP Javelin™ Guard Columns > Particle Size: 5μm; 10L x 2.1mm I.D.
Spot an opportunity for improvement?Share a Content Correction