Learn More
Fisherbrand™ Castaloy™ Chain Clamp
For holding bottles, beakers and other large objects
Brand:  Fisherbrand™ 05745Q
Product Code. 15596700
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
For holding bottles, beakers and other large objects
  • Holds 76 to 165mm (3 to 6.5 in.) bottles and large objects
  • Fast, sure way to secure large equipment to rods and lattices
  • Loop steel chain around object; attach to hook on movable slide; tighten slide with large, nonslip knob
  • Holds equipment firmly in place at any angle, variable distance from support
  • Arm is 15cm (6 in.) long and 13mm (0.5 in.) diameter
  • Use with 05-754Q clamp holder
Specifications
Specifications
1 Ea.
Stainless Steel
For holding bottles, beakers and other large objects
15.24 cm
Chain Clamp
05-754 Q Clamp Holder
6 in.