Learn More
TH1L Mouse anti-Human, CoraLite 488, Clone: 1A10G2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67380-100UL
This item is not returnable.
View return policy
Description
The TH1L antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human TH1L. This antibody recognizes human, mouse, rat antigen. The TH1L antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
TH1L | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human | |
IgG2a |
Immunofluorescence | |
1A10G2 | |
CoraLite 488 | |
100 μL | |
51497 | |
Liquid |