missing translation for 'onlineSavingsMsg'
Learn More

Tetraisopropyltin, 97+%, Thermo Scientific™

Product Code. 11314547
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2949-42-0
Molecular Formula C12H28Sn
Molecular Weight (g/mol) 291.066
MDL Number MFCD00026346
InChI Key NJASUIDIZMMYED-UHFFFAOYSA-N
Synonym tetraisopropyltin, stannane, tetraisopropyl, tetraisopropylstannane, stannane, tetrakis 1-methylethyl, stannane,tetrakis 1-methylethyl, tetra isopropyltin, tetra-i-propyltin, tetraisopropyl-stannane, acmc-1ckip, iso-c3h7 4sn
PubChem CID 18063
IUPAC Name tetra(propan-2-yl)stannane
SMILES CC(C)[Sn](C(C)C)(C(C)C)C(C)C

Specifications

Color Colorless
Density 1.124 g/mL
Boiling Point 112°C to 114°C (18 mmHg)
Odor Characteristic
Linear Formula Sn[(CH3)2CH]4
Refractive Index 1.4851
Quantity 10 g
UN Number UN2788
Sensitivity Air and moisture sensitive
Formula Weight 291.05
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Tetraisopropyltin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.