Learn More
Tetraethylammonium hydroxide, 35% w/w aq. soln., Thermo Scientific Chemicals
69.80 € - 488.00 €
Chemical Identifiers
CAS | 77-98-5 |
---|---|
Molecular Formula | C8H21NO |
Molecular Weight (g/mol) | 147.26 |
MDL Number | MFCD00009024 |
InChI Key | LRGJRHZIDJQFCL-UHFFFAOYSA-M |
Synonym | tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium |
PubChem CID | 6509 |
IUPAC Name | tetraethylazanium;hydroxide |
SMILES | [OH-].CC[N+](CC)(CC)CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11327988
|
Thermo Scientific Alfa Aesar
036560.22 |
100 g |
69.80 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307988
|
Thermo Scientific Alfa Aesar
036560.36 |
500 g |
172.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317988
|
Thermo Scientific Alfa Aesar
036560.A3 |
2 kg |
488.00 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tetraethylammonium hydroxide is a strong organic base and finds applications in photovoltaic chemicals, advanced ceramics, flat panel display chemicals, selective etching and transesterification catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
77-98-5 | |
147.26 | |
LRGJRHZIDJQFCL-UHFFFAOYSA-M | |
6509 | |
[OH-].CC[N+](CC)(CC)CC |
C8H21NO | |
MFCD00009024 | |
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium | |
tetraethylazanium;hydroxide |
Specifications
77-98-5 | |
1.023 g/mL | |
(CH3CH2)4NOH | |
MFCD00009024 | |
14,9201 | |
Miscible with water. | |
[OH-].CC[N+](CC)(CC)CC | |
147.26 | |
147.26 | |
Liquid |
Colorless to Yellow | |
C8H21NO | |
1.404 | |
UN3267 | |
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium | |
LRGJRHZIDJQFCL-UHFFFAOYSA-M | |
tetraethylazanium;hydroxide | |
6509 | |
35% w/w aq. soln. | |
Tetraethylammonium hydroxide |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H300-H311-H314-H335-H370-H372
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 201-073-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only