missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hydrogen sulfate, 98%, Thermo Scientific Chemicals

34.87 € - 578.88 €

Chemical Identifiers

CAS 32503-27-8
Molecular Formula C16H37NO4S
Molecular Weight (g/mol) 339.54
MDL Number MFCD00011637
InChI Key SHFJWMWCIHQNCP-UHFFFAOYSA-M
Synonym tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
PubChem CID 94433
IUPAC Name hydrogen sulfate;tetrabutylazanium
SMILES OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10593911
View Documents
Thermo Scientific Acros
168380250
25 g Plastic bottle
34.87 €
25g
Please to purchase this item. Need a web account? Register with us today!
10743551
View Documents
Thermo Scientific Acros
168381000
100 g Plastic bottle
96.58 €
100g
Please to purchase this item. Need a web account? Register with us today!
10503921
View Documents
Thermo Scientific Acros
168382500
250 g Plastic bottle
217.08 €
250g
Please to purchase this item. Need a web account? Register with us today!
10397820
View Documents
Thermo Scientific Acros
168380010
1 kg Plastic bottle
578.88 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Chemical Identifiers

32503-27-8
339.54
SHFJWMWCIHQNCP-UHFFFAOYSA-M
94433
OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
C16H37NO4S
MFCD00011637
tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
hydrogen sulfate;tetrabutylazanium

Specifications

32503-27-8
White to Beige
98%
[CH3(CH2)3]4NHSO4
06,565; 07,354; 13,94
Solubility in water: >50%.
OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
339.54
339.53
Crystalline Powder and/or Chunks
168.0°C to 172.0°C
Authentic
C16H37NO4S
MFCD00011637
tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
SHFJWMWCIHQNCP-UHFFFAOYSA-M
hydrogen sulfate;tetrabutylazanium
94433
98%
Tetrabutylammonium hydrogen sulfate, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 251-068-5

missing translation for 'tsca' : TSCA

Product Identifier
  • Tetrabutylammonium hydrogen sulfate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.