missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) nitrate, Premion™, 99.99% (metals basis), Pt 50% min, Thermo Scientific Chemicals

96.58 € - 815.06 €

Chemical Identifiers

CAS 20634-12-2
Molecular Formula H12N6O6Pt
Molecular Weight (g/mol) 387.22
MDL Number MFCD00011624
InChI Key RBAKORNXYLGSJB-UHFFFAOYSA-N
Synonym tetraammineplatinum ii nitrate, tetraammineplatinum dinitrate, tetrammineplatinum ii nitrate, tetraammineplatinum ii nitrate 1g, tetraammineplatinum ii nitrate, premion, platinum 2+ tetraamine dinitrate, tetraammineplatinum ii nitrate solution, pt w/w cont. pt
PubChem CID 146543
SMILES N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11314938
SDS View Documents
Thermo Scientific Alfa Aesar
043897.01
0.25 g
96.58 €
0.25g
Please to purchase this item. Need a web account? Register with us today!
11304938
SDS View Documents
Thermo Scientific Alfa Aesar
043897.03
1 g
243.21 €
1g
Please to purchase this item. Need a web account? Register with us today!
11394928
SDS View Documents
Thermo Scientific Alfa Aesar
043897.06
5 g
815.06 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tetraammineplatinum(II) nitrate is widely utilized as a precursor to bimetallic Pt/Zn catalysts for the selective hydrogenation of crotonaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c

H272-H315-H317-H319-H335

missing translation for 'dotInformation' : DOT Class: 5.1; Packaging Group: II

missing translation for 'einecsNumber' : 243-929-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetraammineplatinum(II) nitrate, Premion™, 99.99% (metals basis), Pt 50% min, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.