Learn More
Tetraammineplatinum(II) chloride monohydrate, Premion™, 99.995% (metals basis), Thermo Scientific Chemicals
268.34 € - 833.15 €
Chemical Identifiers
CAS | 13933-33-0 |
---|---|
Molecular Formula | Cl2H14N4OPt |
Molecular Weight (g/mol) | 352.12 |
MDL Number | MFCD00149947 |
InChI Key | GWBDNMCYHWRSOH-UHFFFAOYSA-L |
Synonym | azane;platinum 2+ ;dichloride;hydrate, tetrammineplatinum ii chloride hydrate, tetraammineplatinum ii chloride monohydrate, platinum 2+ ion tetraamine hydrate dichloride, platinum 2+ tetraamine hydrate dichloride, platinum 2+ chloride-ammonia-water 1/2/4/1, tetraamineplatinium ii chloride |
PubChem CID | 21946612 |
IUPAC Name | azane;platinum(2+);dichloride;hydrate |
SMILES | N.N.N.N.O.[Cl-].[Cl-].[Pt++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11376967
|
Thermo Scientific Alfa Aesar
010836.03 |
1 g |
268.34 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386967
|
Thermo Scientific Alfa Aesar
010836.06 |
5 g |
833.15 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tetraammineplatinum(II) chloride is used as a catalyst in the chemical synthesis and a precursor for platinum catalysts. This complex has pharmaceutical applications and used in proteomics research. It is used in the preparation of Pt catalysts supported on reduced graphite oxide (RGO).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13933-33-0 | |
352.12 | |
GWBDNMCYHWRSOH-UHFFFAOYSA-L | |
21946612 | |
N.N.N.N.O.[Cl-].[Cl-].[Pt++] |
Cl2H14N4OPt | |
MFCD00149947 | |
azane;platinum 2+ ;dichloride;hydrate, tetrammineplatinum ii chloride hydrate, tetraammineplatinum ii chloride monohydrate, platinum 2+ ion tetraamine hydrate dichloride, platinum 2+ tetraamine hydrate dichloride, platinum 2+ chloride-ammonia-water 1/2/4/1, tetraamineplatinium ii chloride | |
azane;platinum(2+);dichloride;hydrate |
Specifications
250°C (dec.) | |
White through Cream | |
Cl2H14N4OPt | |
azane;platinum 2+ ;dichloride;hydrate, tetrammineplatinum ii chloride hydrate, tetraammineplatinum ii chloride monohydrate, platinum 2+ ion tetraamine hydrate dichloride, platinum 2+ tetraamine hydrate dichloride, platinum 2+ chloride-ammonia-water 1/2/4/1, tetraamineplatinium ii chloride | |
GWBDNMCYHWRSOH-UHFFFAOYSA-L | |
azane;platinum(2+);dichloride;hydrate | |
21946612 | |
99.995% | |
Hygroscopic | |
(metals basis) |
13933-33-0 | |
Crystalline | |
MFCD00149947 | |
Soluble in water | |
N.N.N.N.O.[Cl-].[Cl-].[Pt++] | |
352.12 | |
352.13 (334.11 Anhydrous) | |
Premion™ | |
2.74g/mL | |
Tetraammineplatinum(II) chloride monohydrate |
Safety and Handling
GHS H Statement
H290-H315-H319
May be corrosive to metals.
Causes skin irritation.
Causes serious eye irritation.
P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
H290-H315-H319
missing translation for 'einecsNumber' : 237-706-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only