missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-hexylammonium benzoate, ca 85%, water ca 10%, Thermo Scientific™

Product Code. 11440878
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16436-29-6
Molecular Formula C31H57NO2
Molecular Weight (g/mol) 475.80
MDL Number MFCD00041982
InChI Key PSEQWFPWQRZBOO-UHFFFAOYSA-M
Synonym tetrahexylammonium benzoate, 1-hexanaminium, n,n,n-trihexyl-, benzoate, tetrahexylazanium benzoate, tetra-n-hexylammonium benzoate, 1-hexanaminium, n,n,n-trihexyl-, benzoate 1:1, tetrahexylazanium, thab, ammonium, tetrahexyl-, benzoate, ammonium, benzoate, tetrahexylazanium,benzoate
PubChem CID 85419
IUPAC Name tetrahexylazanium;benzoate
SMILES [O-]C(=O)C1=CC=CC=C1.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

Specifications

Quantity 5 g
Formula Weight 456.73
Percent Purity ∼85%
Assay water ≈10%
Chemical Name or Material Tetra-n-hexylammonium benzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.