missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium phosphate, 99+%, Thermo Scientific Chemicals

Used as a counter-ion in ion-pair chromatography of water-soluble vitamins

39.95 € - 1115.55 €

Chemical Identifiers

CAS 5574-97-0
Molecular Formula C16H38NO4P
Molecular Weight (g/mol) 339.46
MDL Number MFCD00064526
InChI Key ARRNBPCNZJXHRJ-UHFFFAOYSA-M
Synonym tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
PubChem CID 2735142
IUPAC Name dihydrogen phosphate;tetrabutylazanium
SMILES OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11353517
View Documents
Thermo Scientific Alfa Aesar
019261.02
0.5 g
39.95 €
0.50g
Please to purchase this item. Need a web account? Register with us today!
11363517
View Documents
Thermo Scientific Alfa Aesar
019261.04
2 g
94.57 €
2g
Please to purchase this item. Need a web account? Register with us today!
11343517
View Documents
Thermo Scientific Alfa Aesar
019261.18
50 g
1115.55 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tetra-n-butylammonium phosphate is used as a counter-ion in ion-pair reagent in high-performance liquid chromatography for the simultaneous determination of chlorpheniramine and maleate of water-soluble vitamins. It acts as a phase transfer catalyst. Further, it is used as a buffering reagent in HPLC. In addition to this, it is used as a mobile phase additive in two-dimensional reversed-phase high-performance thin-layer chromatography for the separation of chenodeoxycholic acid and deoxycholic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5574-97-0
339.46
ARRNBPCNZJXHRJ-UHFFFAOYSA-M
2735142
OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
C16H38NO4P
MFCD00064526
tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
dihydrogen phosphate;tetrabutylazanium

Specifications

5574-97-0
White
[CH3(CH2)3]4NH2PO4
5196532
Soluble in acetonitrile,ethanol and water.
OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
339.46
339.46
Crystalline
151°C to 154°C
C16H38NO4P
MFCD00064526
tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
ARRNBPCNZJXHRJ-UHFFFAOYSA-M
dihydrogen phosphate;tetrabutylazanium
2735142
≥99%
Tetra-n-butylammonium phosphate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

H500

missing translation for 'einecsNumber' : 226-947-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.