Learn More
Tetra-n-butylammonium hydroxide, 40% w/w in methanol, Thermo Scientific Chemicals
52.20 € - 657.00 €
Chemical Identifiers
CAS | 2052-49-5 |
---|---|
Molecular Formula | C16H37NO |
Molecular Weight (g/mol) | 259.48 |
MDL Number | MFCD00009425 |
InChI Key | VDZOOKBUILJEDG-UHFFFAOYSA-M |
Synonym | tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 |
PubChem CID | 2723671 |
IUPAC Name | tetrabutylazanium;hydroxide |
SMILES | [OH-].CCCC[N+](CCCC)(CCCC)CCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11480453
|
Thermo Scientific Alfa Aesar
A12626.AE |
100 mL |
52.20 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490453
|
Thermo Scientific Alfa Aesar
A12626.AP |
500 mL |
172.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400463
|
Thermo Scientific Alfa Aesar
A12626.0F |
2500 mL |
657.00 €
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tetra-n-butylammonium hydroxide, 40% w/w in methanol is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is also used as a curing accelerator for epoxy resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetra-n-butylammonium hydroxide, 40% w/w in methanol is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is also used as a curing accelerator for epoxy resins.
Solubility
Miscible with organic solvents.
Notes
Air sensitive and hygroscopic. Incompatible with strong acids and corrodes metal.
Chemical Identifiers
2052-49-5 | |
259.48 | |
VDZOOKBUILJEDG-UHFFFAOYSA-M | |
2723671 | |
[OH-].CCCC[N+](CCCC)(CCCC)CCCC |
C16H37NO | |
MFCD00009425 | |
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 | |
tetrabutylazanium;hydroxide |
Specifications
2052-49-5 | |
18°C (64°F) | |
C16H37NO | |
MFCD00009425 | |
3571228 | |
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 | |
VDZOOKBUILJEDG-UHFFFAOYSA-M | |
tetrabutylazanium;hydroxide | |
2723671 | |
40% w/w in methanol |
0.91 | |
Alcohol-like | |
1.4 | |
UN2924 | |
Air sensitive; Hygroscopic | |
Miscible with organic solvents. | |
[OH-].CCCC[N+](CCCC)(CCCC)CCCC | |
259.48 | |
259.48 | |
Tetra-n-butylammonium hydroxide |
Safety and Handling
GHS H Statement
H225-H301-H311-H331-H370-H314-H318
Highly flammable liquid and vapour.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H301+H311+H331-H314-H335-H370
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 218-147-6
missing translation for 'rtecsNumber' : BS5425000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only