Learn More
Tetra-n-butylammonium fluoride, 1M soln. in THF, Thermo Scientific Chemicals
45.00 € - 429.00 €
Chemical Identifiers
CAS | 429-41-4 |
---|---|
Molecular Formula | C16H36FN |
Molecular Weight (g/mol) | 261.47 |
MDL Number | MFCD00011747 |
InChI Key | FPGGTKZVZWFYPV-UHFFFAOYSA-M |
Synonym | tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1 |
PubChem CID | 2724141 |
ChEBI | CHEBI:51990 |
IUPAC Name | tetrabutylazanium;fluoride |
SMILES | [F-].CCCC[N+](CCCC)(CCCC)CCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11443940
|
Thermo Scientific Alfa Aesar
A10588.AC |
25 mL |
45.00 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433254
|
Thermo Scientific Alfa Aesar
A10588.AE |
100 mL |
112.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11443254
|
Thermo Scientific Alfa Aesar
A10588.AP |
500 mL |
429.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetra-n-butylammonium fluoride, 1M soln. in THF is used as a reactant for the preparation of triple monoamine reuptake inhibitors as a new generation of antidepressants, oligoribonucleotides with phosphonate-modified linkages, conjugated dienoic acid esters using Suzuki coupling reactions and polyacetylenic glucosides. It plays an important role in aryl alkyl alcohols through Nozaki-Hiyama allylation catalyzed by chiral bipyridyldiol ligands and chromium trichloride. It is also used as a catalyst for the silylation reactions. In addition, it is used as a source of fluoride ion in organic solvents as well as for the deprotection of silyl ether groups.
Solubility
Miscible with terahydrofuran, acetonitrile, dimethyl sulfoxide and organic solvents.
Notes
Store in a cool place. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and oxygen.
Chemical Identifiers
429-41-4 | |
261.47 | |
FPGGTKZVZWFYPV-UHFFFAOYSA-M | |
2724141 | |
tetrabutylazanium;fluoride |
C16H36FN | |
MFCD00011747 | |
tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1 | |
CHEBI:51990 | |
[F-].CCCC[N+](CCCC)(CCCC)CCCC |
Specifications
429-41-4 | |
−17°C (1°F) | |
[CH3(CH2)3]4NF | |
UN2924 | |
Hygroscopic | |
Miscible with terahydrofuran,acetonitrile,dimethyl sulfoxide and organic solvents. | |
[F-].CCCC[N+](CCCC)(CCCC)CCCC | |
261.47 | |
CHEBI:51990 | |
1M soln. in THF |
0.903 | |
C16H36FN | |
MFCD00011747 | |
3570522 | |
tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1 | |
FPGGTKZVZWFYPV-UHFFFAOYSA-M | |
tetrabutylazanium;fluoride | |
2724141 | |
261.47 | |
Tetra-n-butylammonium fluoride |
Safety and Handling
GHS H Statement
H225-H314-H351-H302-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335-H336-H351-H373
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 207-057-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only