missing translation for 'onlineSavingsMsg'
Learn More

Tetra-N-Butyl Ammonium Hydroxide Solution 0.1M (0.1N) In Toluene/Methanol, Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

166.00 € - 1110.00 €

Chemical Identifiers

CAS 2052-49-5
Molecular Formula C16H37NO
Molecular Weight (g/mol) 259.48
MDL Number MFCD00009425
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
PubChem CID 2723671
IUPAC Name tetrabutylazanium hydroxide
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10588050
View Documents
Fisher Chemical
J/8750/08
500 mL Amber glass bottle
166.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10305810
View Documents
Fisher Chemical
J/8750/15
1 L Amber glass bottle
334.00 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10152100
View Documents
Fisher Chemical
J/8750/17
2.5 L Amber glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11408972
View Documents
Fisher Chemical
J/8750/24
10 L Bag in box
1110.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tetra-N-Butyl Ammonium Hydroxide Solution 0.1M (0.1N) In Toluene/Methanol, Standard Solution Ready To Use, for Volumetric Analysis

Specifications

Chemical Identifiers

2052-49-5
259.48
VDZOOKBUILJEDG-UHFFFAOYSA-M
2723671
[OH-].CCCC[N+](CCCC)(CCCC)CCCC
C16H37NO
MFCD00009425
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
tetrabutylazanium hydroxide

Specifications

2052-49-5
Liquid
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylazanium hydroxide
2723671
Colorless
C16H37NO
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
[OH-].CCCC[N+](CCCC)(CCCC)CCCC
259.48
259.47g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H370-Causes damage to organs.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.