Learn More
Lenz Laborglasintrumente Test Tubes with NS Joint, with Graduation
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05706004
Product Code. 11973571
  • 298.00 € / Pack of 10
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With round bottom
  • With NS socket
  • With PE stopper
  • With scale in high-contrast enamel
  • Made of DURAN™ tubing
Specifications
Specifications
30 mL
Borosilicate Glass (3.3)