missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals

213.00 € - 879.00 €

Chemical Identifiers

CAS 677-22-5
Molecular Formula C4H9ClMg
Molecular Weight (g/mol) 116.87
MDL Number MFCD00000465
InChI Key ZDRJSYVHDMFHSC-UHFFFAOYSA-M
Synonym tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgcl
PubChem CID 2724198
IUPAC Name magnesium;2-methylpropane;chloride
SMILES CC(C)(C)[Mg]Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15458047
View Documents
Thermo Scientific Alfa Aesar
H51164.AE
100 mL
213.00 €
100mL
Please to purchase this item. Need a web account? Register with us today!
15468047
View Documents
Thermo Scientific Alfa Aesar
H51164.AP
500 mL
879.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

tert-Butylmagnesium chloride is a Grignard reagent used in organic synthesis. It is also involved in copper-catalyzed cross-coupling reaction with primary-alkyl halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butylmagnesium chloride is a Grignard reagent used in organic synthesis. It is also involved in copper-catalyzed cross-coupling reaction with primary-alkyl halides.

Solubility
Miscible with alcohol and water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and bases.
Specifications

Chemical Identifiers

677-22-5
116.87
ZDRJSYVHDMFHSC-UHFFFAOYSA-M
2724198
CC(C)(C)[Mg]Cl
C4H9ClMg
MFCD00000465
tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgcl
magnesium;2-methylpropane;chloride

Specifications

677-22-5
(CH3)3CMgCl
UN3399
tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgcl
ZDRJSYVHDMFHSC-UHFFFAOYSA-M
magnesium;2-methylpropane;chloride
2724198
1M in MeTHF
C4H9ClMg
MFCD00000465
Air and moisture sensitive
Miscible with alcohol and water.
CC(C)(C)[Mg]Cl
116.87
116.87
tert-Butylmagnesium chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H261-H314
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.

P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c

H225-H260-H302-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.