Learn More
tert-Butyldimethylchlorosilane, 50% w/w in toluene, Thermo Scientific Chemicals
121.61 € - 1073.00 €
Chemical Identifiers
CAS | 18162-48-6 |
---|---|
Molecular Formula | C6H15ClSi |
Molecular Weight (g/mol) | 150.721 |
MDL Number | MFCD00000501 |
InChI Key | BCNZYOJHNLTNEZ-UHFFFAOYSA-N |
Synonym | tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride |
PubChem CID | 28928 |
ChEBI | CHEBI:85071 |
IUPAC Name | tert-butyl-chloro-dimethylsilane |
SMILES | CC(C)(C)[Si](C)(C)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11409197
|
Thermo Scientific Alfa Aesar
B21286.18 |
50 g |
121.61 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419197
|
Thermo Scientific Alfa Aesar
B21286.30 |
250 g | N/A | ||||||
11499187
|
Thermo Scientific Alfa Aesar
B21286.0B |
1000 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18162-48-6 | |
150.721 | |
BCNZYOJHNLTNEZ-UHFFFAOYSA-N | |
28928 | |
tert-butyl-chloro-dimethylsilane |
C6H15ClSi | |
MFCD00000501 | |
tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride | |
CHEBI:85071 | |
CC(C)(C)[Si](C)(C)Cl |
Specifications
18162-48-6 | |
(CH3)3CSi(CH3)2Cl | |
UN2924 | |
tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride | |
CC(C)(C)[Si](C)(C)Cl | |
150.721 | |
CHEBI:85071 | |
50% w/w in toluene | |
Moisture sensitive | |
tert-Butyldimethylchlorosilane |
C6H15ClSi | |
MFCD00000501 | |
7°C (45°F) | |
BCNZYOJHNLTNEZ-UHFFFAOYSA-N | |
tert-butyl-chloro-dimethylsilane | |
28928 | |
150.73 | |
Characteristic | |
0.87 |
Safety and Handling
GHS H Statement
H225-H314-H318-H361-H373-H336
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H304-H314-H335-H336-H361d-H373
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- tert-Butyldimethylchlorosilane, 50% w/w in toluene
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H336-May cause drowsiness or dizziness.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Toluene, tert-Butyldimethylchlorosilane
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.