missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl methyl ether, For pesticide residue analysis, Honeywell Riedel-de Haën™
CHROMASOLV™, for pesticide residue analysis
82.90 € - 594.63 €
Chemical Identifiers
CAS | 1634-04-4 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
MDL Number | MFCD00008812 |
InChI Key | BZLVMXJERCGZMT-UHFFFAOYSA-N |
Synonym | tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane |
PubChem CID | 15413 |
ChEBI | CHEBI:27642 |
IUPAC Name | 2-methoxy-2-methylpropane |
SMILES | CC(C)(C)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15644230
|
Honeywell-Riedel-de Haen
34498-1L |
1 L |
82.90 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15654230
|
Honeywell-Riedel-de Haen
34498-2.5L |
2.5 L |
170.36 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15664230
|
Honeywell-Riedel-de Haen
34498-4X2.5L |
4 x 2.5 L |
594.63 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Chemical Identifiers
1634-04-4 | |
88.15 | |
BZLVMXJERCGZMT-UHFFFAOYSA-N | |
15413 | |
2-methoxy-2-methylpropane |
C5H12O | |
MFCD00008812 | |
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
CHEBI:27642 | |
CC(C)(C)OC |
Specifications
1634-04-4 | |
0.151 | |
705°F | |
(CH3)3COCH3 | |
UN2398 | |
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
CC(C)(C)OC | |
88.15 | |
CHEBI:27642 | |
≥99.7% | |
tert-Butyl methyl ether |
0.74g/mL at 25°C | |
−33°C (−27.4°F) | |
C5H12O | |
MFCD00008812 | |
1730942 | |
BZLVMXJERCGZMT-UHFFFAOYSA-N | |
2-methoxy-2-methylpropane | |
15413 | |
88.15g/mol | |
CHROMASOLV™ |
Safety and Handling
P210
missing translation for 'rtecsNumber' : KN5250000