missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™

CHROMASOLV™, for HPLC,≥99.8%

65.04 € - 902.33 €

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15654660
View Documents
Honeywell-Riedel-de Haen
34875-100ML
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15664660
View Documents
Honeywell-Riedel-de Haen
34875-1L
1 L
135.74 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15684660
View Documents
Honeywell-Riedel-de Haen
34875-2L
2 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15674660
View Documents
Honeywell-Riedel-de Haen
34875-2.5L
2.5 L
258.07 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15604670
View Documents
Honeywell-Riedel-de Haen
34875-6X1L
6 x 1 L
585.81 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15694660
View Documents
Honeywell-Riedel-de Haen
34875-4X2.5L
4 x 2.5 L
902.33 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
Specifications

Chemical Identifiers

1634-04-4
88.15
BZLVMXJERCGZMT-UHFFFAOYSA-N
CHEBI:27642
C5H12O
MFCD00008812
15413
2-methoxy-2-methylpropane

Specifications

1634-04-4
55°C to 56°C
C5H12O
MFCD00008812
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
CC(C)(C)OC
88.15
CHEBI:27642
≥99.8%
tert-Butyl methyl ether, for HPLC
0.74g/mL at 25°C
n20/D 1.369
(CH3)3COCH3
1730942
BZLVMXJERCGZMT-UHFFFAOYSA-N
2-methoxy-2-methylpropane
15413
88.15g/mol
CHROMASOLV™
Videos
Safety and Handling

Safety and Handling

P210

H225-H315

missing translation for 'einecsNumber' : 216-653-1

missing translation for 'rtecsNumber' : KN5250000

Product Identifier
  • tert-Butyl methyl ether

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.

Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.