Learn More
Dostmann Electronic™ Temperature Sensor, Thermocouple
Dostmann Electronic™ Temperature Sensor, Thermocouple is a plug and cable type temperature sensor suitable for variety of temperature measurements demands.
Brand:  Dostmann Electronic™ 6010-0058
 View more versions of this product
Product Code. 10725183
  • 28.35 € / Each
Explore more special offers
Description
Description
  • Type K according to EN 60584-1
  • NiCr-Ni
  • Mineral-insulated
Specifications
Specifications
1 mm
Temperature Sensor
Up to 400°C
1000 mm
Thermocouple, fiberglass insulated
P410 / TC301 / TC 309 / Mini-K