Learn More
Dostmann Electronic™ Temperature Humidity Instrument
This robust, reliable, ready-to-use temperature humidity instrument is a perfectly designed instrument for climatic measurements (HVAC branch).
Brand:  Dostmann Electronic™ 5020-0560
 View more versions of this product
Product Code. 10543113
  • 116.00 € / Each
Explore more special offers
Description
Description
  • Perfectly designed instrument for climatic meas­ure­ments (HVAC branch). 
    • The measuring instruments are perfect for measuring room climate (HVAC)
    • For detecting critical temperature-humidity environments: moul-risk in living quarters
    • The flexible combination sensor of P330 is excellent useable for measurements at hard-to-access points
Specifications
Specifications
Thermohygrometer
±3%
±0.5°C
Plastic
130 g
0 to 100%
-40°C to +70°C
LCD
170 x 50 x 17 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products