Learn More
Cole-Parmer™ CB-200D Block Chiller/Heater
Description
- Designed for maintaining biological samples safely between 4 and 37°C
- Alarm sounds when set temperature is reached and elapsed time ends
- Five-digit LED display can be set for either °C or °F
- 30 min. Cool Down Time to 4°C
- Choose from a wide range of interchangeable aluminum block inserts
- 3 year warranty
- Previous part number FBLOCKICE
This benchtop block chiller/heater is ideal for incubating sample tubes at temperatures ranging from 32 to 104°F (0 to 40°C). The Isotemp™ temperature control ensures all contents are kept at an even temperature, whether cold or warm. Integrated countdown/count-up timer with audible alarm allows up to nine days to be programmed and will not start until a stable temperature has been reached.
Use with Dri-Block™ aluminum inserts to maintain temperature stability of ±0.2°C. Unit holds two single-width inserts or one double-width inserts. A dark plastic lid protects light-sensitive reagents as well as helping to maintain the sample temperature.
Please Note: Effective June 2022, the Techne Cooling/Heating Block Model number FBLOCKICE has now been rebranded Cole-Parmer under Model number CB-200D. All the same great features and specifications, just with a new and improved look.
Specifications
Specifications
For Use With (Equipment) | Aluminum Blocks |
Temperature Range (Metric) | 0°C to 40°C |
Type | Block |
Dimensions (L x W x H) | 190 x 240 x 225 mm |
Length (English) | 7.5 in. |
Length (Metric) | 190 mm |
Width (English) | 9.4 in. |
Width (Metric) | 240 mm |
Height (English) | 8.9 in. |
Height (Metric) | 225 mm |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.