Learn More
TCR Beta Armenian Hamster anti-Mouse, APC, Clone: H57-597, Proteintech
Armenian Hamster Monoclonal Antibody
Brand: Proteintech APC-65106-100UG
This item is not returnable.
View return policy
Description
The TCR Beta antibody from Proteintech is a armenian hamster monoclonal antibody generated with affinity purified tcr from mouse do-11.10 cells of mouse TCR Beta. This antibody recognizes mouse antigen. The TCR Beta antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
TCR Beta | |
Monoclonal | |
0.2 mg/mL | |
Armenian Hamster | |
Primary | |
Mouse |
Flow Cytometry | |
H57-597 | |
APC | |
100 μg | |
21577 | |
IgG |