Learn More
Thermo Scientific Chemicals Taurocholic acid, sodium salt hydrate, 98%
Taurocholic acid, sodium salt hydrate, CAS # 312693-83-7, also known as sodium taurocholate hydrate, is a bile acid taurine conjugate form in the liver with detergent capabilities.
54.47 € - 2089.40 €
Chemical Identifiers
CAS | 345909-26-4 |
---|---|
Molecular Formula | C26H44NNaO7S |
Molecular Weight (g/mol) | 537.69 |
MDL Number | MFCD00150819 |
InChI Key | JAJWGJBVLPIOOH-IZYKLYLVSA-M |
Synonym | Sodium taurocholate hydrate |
PubChem CID | 23687511 |
SMILES | [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC(=O)NCCS([O-])(=O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10701701
|
Thermo Scientific Chemicals
230370010 |
1 g | Glass bottle |
54.47 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10617012
|
Thermo Scientific Chemicals
230370050 |
5 g | Glass bottle |
172.86 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10629453
|
Thermo Scientific Chemicals
230370250 |
25 g | Glass bottle |
738.68 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11995541
|
Thermo Scientific Chemicals
230371000 |
100 g | Glass bottle |
2089.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Taurocholic acid, sodium salt hydrate is an anionic bile acid detergent synthesized from cholic acid
- This compound shows a choleretic activity promoting the secretion of bile from the liver
Applications
- Taurocholic acid is used in the emulsification of lipids/ fats
- It is used for protein solubilization and in the preparation of digestive enzymes for research applications
- It can be used to determine conjugated and unconjugated bile salts in serum and jejunal fluid
Chemical Identifiers
345909-26-4 | |
537.69 | |
JAJWGJBVLPIOOH-IZYKLYLVSA-M | |
23687511 |
C26H44NNaO7S | |
MFCD00150819 | |
Sodium taurocholate hydrate | |
[Na+].[H][C@@]12CC[C@H]([C@H](C)CCC(=O)NCCS([O-])(=O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C |
Specifications
Undesignated | |
Powder | |
98% | |
C26H44NNaO7S | |
MFCD00150819 | |
JAJWGJBVLPIOOH-IZYKLYLVSA-M | |
sodium 2-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]ethane-1-sulfonate | |
23687511 | |
≥97.5% (TLC) |
345909-26-4 | |
Authentic | |
+20.0° to +25.8° (20°C, 589nm) (c=3, water) | |
7% max. | |
Sodium taurocholate hydrate | |
[Na+].[H][C@@]12CC[C@H]([C@H](C)CCC(=O)NCCS([O-])(=O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C | |
537.69 | |
537.7 | |
Taurocholic acid, sodium salt hydrate |
Safety and Handling
GHS H Statement
Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning
missing translation for 'einecsNumber' : 205-653-7
missing translation for 'rtecsNumber' : WX0400000
missing translation for 'tsca' : TSCA
RUO – Research Use Only