missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals TAPS, 99+%, for biochemistry
32.96 € - 74.97 €
Chemical Identifiers
CAS | 29915-38-6 |
---|---|
Molecular Formula | C7H17NO6S |
Molecular Weight (g/mol) | 243.27 |
MDL Number | MFCD00007538 |
InChI Key | YNLCVAQJIKOXER-UHFFFAOYSA-N |
Synonym | 3-[Tris(hydroxymethyl)methylamino]-1-propanesulfonic acid |
PubChem CID | 121591 |
IUPAC Name | 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid |
SMILES | OCC(CO)(CO)NCCCS(O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10716623
|
Thermo Scientific Chemicals
327800250 |
25 g | Glass bottle |
32.96 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10317901
|
Thermo Scientific Chemicals
327801000 |
100 g | Glass bottle |
74.97 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
29915-38-6 | |
243.27 | |
YNLCVAQJIKOXER-UHFFFAOYSA-N | |
121591 | |
OCC(CO)(CO)NCCCS(O)(=O)=O |
C7H17NO6S | |
MFCD00007538 | |
3-[Tris(hydroxymethyl)methylamino]-1-propanesulfonic acid | |
3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid |
Specifications
(0.1 M aq. soln.) | |
29915-38-6 | |
White | |
Powder | |
99+% | |
C7H17NO6S | |
MFCD00007538 | |
3-[Tris(hydroxymethyl)methylamino]-1-propanesulfonic acid | |
YNLCVAQJIKOXER-UHFFFAOYSA-N | |
3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid | |
121591 | |
≥99% | |
TAPS |
(0.1M aq. soln.) | |
240.0°C | |
5 to 6.5 (1% aq. soln. at 25°C) | |
>110°C | |
Authentic | |
(HOCH2)3CNH(CH2)3SO3H | |
15, 9191 | |
Solubility in water: ca 121.6g/L (20°C). | |
OCC(CO)(CO)NCCCS(O)(=O)=O | |
243.27 | |
243.27 | |
Biochemical |
Safety and Handling
Product Identifier
- TAPS
RUO – Research Use Only