Learn More
Fisherbrand™ Tapered PTFE Stir Bars
Provides similar action as oval or pivot ring stirrers. Cowie Technology™ Tapered PTFE Stir Bars are stable and autoclavable for a wide variety of uses.
Brand:  Fisherbrand™ 001.1940
 View more versions of this product
Product Code. 16215771
  • 17.10 € / Pack of 10
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stable, general-purpose magnetic stir bar
  • Tapered ends provide similar action as oval or pivot ring stirrers
  • Autoclavable
Specifications
Specifications
Autoclavable
White
Containers & Vessels / Stirrer Hot Plates
40 mm
Tapered Cylindrical
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
8 mm
5/Pk.