missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micronova™ Tape matt finish, write-on, controlled environment, vinyl, dark blue 50mm x 33m Premium
Brand: Micronova™ CR100PC-2DB
This item is not returnable.
View return policy
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
039697-000 | Certificate of Compliance | 24/08/2023 | CR100PC-2DB |
038788-000 | Certificate of Compliance | 28/06/2023 | CR100PC-2DB |
038787-000 | Certificate of Compliance | 28/06/2023 | CR100PC-2DB |
038328-000 | Certificate of Compliance | 31/05/2023 | CR100PC-2DB |
038214-000 | Certificate of Compliance | 11/05/2023 | CR100PC-2DB |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Micronova™ Tape matt finish, write-on, controlled environment, vinyl, dark blue 50mm x 33m Premium >
Spot an opportunity for improvement?Share a Content Correction