missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micronova™ Tape general purpose polyethylene transparent 19mm x 33m
Industrial grade polyethylene tape for cleanroom support areas, maintenance and clean build projects where ultra-clean tape is not required. Available in black, red, dark blue, yellow, brown, orange, dark green, white and transparent.
Brand: Micronova™ EGPFP-34TR
This item is not returnable.
View return policy
Description
- Polyethylene tape
- For use in areas where ultra-clean tape is not required
- Acrylic adhesive
- Good clean-peel properties
- Various widths and lengths available
Specifications
Transparent | |
For general purpose use | |
33 m |
Polyethylene | |
33 m x 19 mm | |
19 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction