missing translation for 'onlineSavingsMsg'
Learn More
Tantalum(V) oxide, Puratronic™, 99.993% (metals basis excluding Nb), Nb 50ppm max, Thermo Scientific Chemicals Product Code.: 11327057

Tantalum(V) oxide, Puratronic™, 99.993% (metals basis excluding Nb), Nb 50ppm max, Thermo Scientific Chemicals

Product Code. 11327057
100 g
Click to view available options
Quantity:
25 g
100 g
500 g

Product Code. 11327057

Brand: Thermo Scientific Alfa Aesar 010881.22

Please to purchase this item. Need a web account? Register with us today!

It is used in the production of capacitors, due to its high dielectric constant. These capacitors are used in automotive electronics, cell phones, thin film components, and electronic circuitry. In addition, Tantalum(V) oxide has been utilized in the fabrication of the glass of photographic lenses. Due to its high biocompatibility and resistivity, it has attracted many biomedical applications. It is used in tantalum capacitors, resistive switching memories, high frequency CMOS (Complementary metal-oxide-semiconductors) integrated circuits, and high-k dielectric for DRAM (Dynamic random-access memory) capacitor applications. It has been shown to improve corrosion resistance and cytocompatibility of titanium substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-61-0
Molecular Formula O5Ta2
Molecular Weight (g/mol) 441.89
MDL Number MFCD00011254
InChI Key PBCFLUZVCVVTBY-UHFFFAOYSA-N
IUPAC Name [(dioxotantalio)oxy]dioxotantalum
SMILES O=[Ta](=O)O[Ta](=O)=O

Specifications

Melting Point 1800°C
Physical Form Powder
Quantity 100 g
Merck Index 14,9056
Solubility Information Insoluble in almost all organic solvents. Solubility is negligible in water.
Formula Weight 441.89
Percent Purity 99.993%
Grade Puratronic™
Odor Odorless
Density 8.2 g/mL
Assay (metals basis excluding Nb), Nb 50ppm max.
Chemical Name or Material Tantalum(V) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tantalum(V) oxide, Puratronic™, 99.993% (metals basis excluding Nb), Nb 50ppm max, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.