missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tantalum gauze, 80 mesh woven from 0.08mm (0.003 in.) dia. wire, Thermo Scientific Chemicals
117.59 €
Chemical Identifiers
CAS | 7440-25-7 |
---|---|
Molecular Formula | Ta |
Molecular Weight (g/mol) | 180.95 |
MDL Number | MFCD00011252 |
InChI Key | GUVRBAGPIYLISA-UHFFFAOYSA-N |
Synonym | tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod |
PubChem CID | 23956 |
ChEBI | CHEBI:33348 |
IUPAC Name | tantalum |
SMILES | [Ta] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-25-7 | |
180.95 | |
GUVRBAGPIYLISA-UHFFFAOYSA-N | |
23956 | |
tantalum |
Ta | |
MFCD00011252 | |
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
CHEBI:33348 | |
[Ta] |
Specifications
7440-25-7 | |
MFCD00011252 | |
GUVRBAGPIYLISA-UHFFFAOYSA-N | |
tantalum | |
23956 | |
80 Mesh | |
Wire Cloth |
Ta | |
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
[Ta] | |
180.95 | |
CHEBI:33348 | |
Odorless | |
Tantalum gauze, 80 mesh woven from 0.08mm (0.003 in.) dia. wire |
Safety and Handling
missing translation for 'einecsNumber' : 231-13505
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only