Learn More
Tantalum carbide, 99.5% (metals basis), Thermo Scientific Chemicals
74.47 € - 925.61 €
Chemical Identifiers
CAS | 12070-06-3 |
---|---|
Molecular Formula | CTa |
Molecular Weight (g/mol) | 192.96 |
MDL Number | MFCD00011255 |
InChI Key | DUMHRFXBHXIRTD-UHFFFAOYSA-N |
IUPAC Name | methanidylidynetantalumylium |
SMILES | [C-]#[Ta+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11396508
|
Thermo Scientific Alfa Aesar
012144.09 |
10 g |
74.47 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306518
|
Thermo Scientific Alfa Aesar
012144.18 |
50 g |
243.21 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386508
|
Thermo Scientific Alfa Aesar
012144.30 |
250 g |
925.61 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tantalum carbides is used in tungsten carbide. It is also used as a coating for steel moulds in the injection moulding of aluminum alloys. While providing a hard, wear resistant surface, it also provides a low friction mould surface. It is also used in the production of sharp instruments with extreme mechanical resistance and hardness, tool bits for cutting tools.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12070-06-3 | |
192.96 | |
DUMHRFXBHXIRTD-UHFFFAOYSA-N | |
[C-]#[Ta+] |
CTa | |
MFCD00011255 | |
methanidylidynetantalumylium |
Specifications
5500°C | |
12070-06-3 | |
Powder | |
MFCD00011255 | |
Soluble in hydrofluoric acid and nitric acid. Partially soluble in water. | |
[C-]#[Ta+] | |
192.96 | |
99.5% | |
13.9 g/mL | |
Tantalum carbide |
3880°C | |
Black to Gray | |
CTa | |
UN3178 | |
DUMHRFXBHXIRTD-UHFFFAOYSA-N | |
methanidylidynetantalumylium | |
192.96 | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 235-118-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only